Level A Suit Maintenance
fisher-scientific-safety-services-level-a-suits-banner-desktop
Overview
Industry standards recommend that a visual inspection and pressure testing of Level A ensembles be done at the following times:
  • Upon receipt from supplier
  • At least once per year
  • After the garment is worn and before the garment is worn again
Note: Contaminated, damaged or altered garments should not be re-used.
Services We Offer
  • Annual pressure testing (using "pass/fail" parameter; no repairs)
  • Pressure testing after each use ("pass/fail" parameter; no repairs)
Note: All suits must be decontaminated before testing at our service centers is performed Fisher Scientific will provide certification form. All NFPA Level A suits should be tested according to ASTM F 1052, Standard Test Method for Pressure Testing Vapor Protective Ensembles, 29CFR1910.120, Hazardous Waste Operations and Emergency Response (HAZWOPER) and EN 464, Determination of Leaktightness of Gas-tight Suits.
Brands We Service
Fisher Scientific Safety Services
Speciality Programs
U.S. Navy
Remove the worry with our convenient onsite service.
Learn More
Concierge Program
We'll guide you to the solution that best fits your needs.
Learn More
Contact Us
Our call center is open Monday-Friday, 8:00 a.m. to 5:00 p.m. EST. +1 866-822-2723 or contact the service team.