Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB780 Mouse Anti-C-Peptide, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570458
Description
C-Peptide Monoclonal antibody specifically detects C-Peptide in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
C-Peptide | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
U8-424 | |
RB780 | |
INS; insulin; preproinsulin; proinsulin | |
Human C-Peptide | |
25 μg | |
3630, 16334, 24506 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction