Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB780 Mouse Anti-C-Peptide, BD Horizon™
Mouse Monoclonal Antibody
$194.00 - $448.00
Specifications
Antigen | C-Peptide |
---|---|
Clone | U8-424 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
C-Peptide Monoclonal antibody specifically detects C-Peptide in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
C-Peptide | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
3630, 16334, 24506 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
U8-424 | |
Flow Cytometry | |
RB780 | |
Mouse | |
Human | |
INS; insulin; preproinsulin; proinsulin | |
Human C-Peptide | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title