Learn More
ELiTech Molecular Diagnostics Real-Time PCR Internal Controls
Specifications
Content And Storage | Chemically synthesized DNA oligonucleotide primers and probes |
---|---|
For Use With (Application) | Real Time PCR (qPCR) |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
22-801-062
|
Nanogen, Inc
800427 |
EBV Internal Controls | EBV Internal Controls | N/A | N/A | N/A | |||
22-801-064
|
Nanogen, Inc
800429 |
HHV-6 Internal Controls | HHV-6 Internal Controls | N/A | N/A | N/A | |||
22-801-065
|
Nanogen, Inc
800430 |
BK Virus Internal Controls | BK Virus Internal Controls | N/A | N/A | N/A | |||
22-801-066
|
Nanogen, Inc
800431 |
CMV 3.0 Internal Controls | CMV 3.0 Internal Controls | N/A | N/A | N/A | |||
22-801-069
|
Nanogen, Inc
800432 |
CMV 3.0 Internal Controls | CMV 3.0 Internal Controls | N/A | N/A | N/A | |||
22-801-072
|
Nanogen, Inc
800439 |
Internal Control Set IC1-V | Internal Control Set IC1-V | N/A | N/A | N/A | |||
Description
Available as FDA-cleared unassayed quality control materials, Real-Time PCR Internal Controls detect variation that may arise during the DNA or RNA extraction and amplification process associated with real-time PCR assays. Each set includes primers and probes that can be used to amplify and detect an internal control template (also included).
All Internal Controls use innovative MGB™ technology, including proprietary probe technology, providing labs with a powerful tool to detect variation in their real-time PCR assays. Real-Time PCR Internal Controls are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into an existing workflow.
- Universal Cycling Conditions: Run multiple assays simultaneously
- Reduced Royalties: Real-time PCR without TaqMan™ probes
- Streamlined Workflow: Bring new molecular tests on quickly
- Open Platform: Runs on most real-time PCR instruments
Specifications
Chemically synthesized DNA oligonucleotide primers and probes | |
Real Time PCR (qPCR) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.