Learn More
ELiTech Molecular Diagnostics Real-Time PCR Internal Controls
Supplier:  Nanogen, Inc 800430
 View more versions of this product
Catalog No. 22-801-065
Description
Description
Available as FDA-cleared unassayed quality control materials, Real-Time PCR Internal Controls detect variation that may arise during the DNA or RNA extraction and amplification process associated with real-time PCR assays. Each set includes primers and probes that can be used to amplify and detect an internal control template (also included).
All Internal Controls use innovative MGB™ technology, including proprietary probe technology, providing labs with a powerful tool to detect variation in their real-time PCR assays. Real-Time PCR Internal Controls are optimized for use with most commercially available real-time PCR instruments. With universal cycling conditions, the reagents can easily be incorporated into an existing workflow.
  • Universal Cycling Conditions: Run multiple assays simultaneously
  • Reduced Royalties: Real-time PCR without TaqMan™ probes
  • Streamlined Workflow: Bring new molecular tests on quickly
  • Open Platform: Runs on most real-time PCR instruments
Specifications
Specifications
Chemically synthesized DNA oligonucleotide primers and probes
BK Virus Internal Control Set
48
Real Time PCR (qPCR)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.