Learn More
DWK Life Sciences Wheaton™ Hybridization Bottles
For use in nucleic acid and protein blotting procedures that require a minimum of probe solution
Supplier:  DWK Life Sciences 805020
 View more versions of this product
Catalog No. 13-245-97F
Description
Description
  • Manufactured from heavy-wall borosilicate glass
  • Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
  • Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
  • Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage
Order Info
Items come as one per case
Specifications
Specifications
35 mm
1.37 in.
75 mm
2.95 in.
Nucleic acid and protein blotting procedures that require a minimum of probe solution
Screw
Polycarbonate
Borosilicate Glass
Safety Coated Hybridization Bottle
45 mm
PTFE or Silicone
USP Type I
Plastisol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.