Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water Solution, For HPLC, Contains 0.1% (v/v) Formic Acid, MilliporeSigma™ Supelco™

The product is water containing 0.1% v/v formic acid. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. It can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques.

$695.00 - $4488.00

Specifications

Linear Formula H2O
MDL Number MFCD00011332
Grade For HPLC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
11-100-3684
View Documents
MilliporeSigma Supelco
5769134X4L
4 x 4 L Pure-Pac™ 1
Each for $695.00
Add to Cart
 
11-100-5322
View Documents
MilliporeSigma Supelco
576913200LLS
200 L Pure-Pac™ 1
Each for $4,488.00
Add to Cart
 
11-100-5323
View Documents
MilliporeSigma Supelco
576913200LP2LSSB
200 L Pure-Pac™ 2 N/A N/A N/A
Description

Description

Specifications

Specifications

H2O
MFCD00011332
For HPLC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.