Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Water Solution, For HPLC, Contains 0.1% (v/v) Formic Acid, MilliporeSigma™ Supelco™
The product is water containing 0.1% v/v formic acid. This pre-blended mobile phase solvent is of high purity that reduces time-consuming preparation and prevents instrument downtime because of impure mobile phases. It can be employed for HPLC and UHPLC (Ultra High performance Liquid Chromatography) techniques.
Supplier: MilliporeSigma Supelco 576913200LP2LSSB
Specifications
H2O | |
Pure-Pac™ 2 | |
For HPLC |
MFCD00011332 | |
200 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction