Learn More
DuPont™ Tychem™ 4000 Coverall with Standard Fit Hood, Attached Socks, Berry Compliant
DuPont™ TYCHEM™ 4000 Coveralls provide at least 30 minutes of protection against >124 chemical challenges in a range of chemical environments.
$554.00 - $728.00
Specifications
Color | White |
---|---|
Disposable | Yes |
Ankle Style | Attached Socks |
Cuff Style | Elastic |
Neck Style | Hooded |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
17-500-145
|
DuPont™
SL122BWH3X0012BN |
3X-Large |
Case of 12 for $554.00
|
|
|||||
17-500-146
|
DuPont™
SL122BWH4X0012BN |
4X-Large |
Case of 12 for $598.00
|
|
|||||
17-500-148
|
DuPont™
SL122BWHLG0012BN |
Large |
Case of 12 for $671.00
|
|
|||||
19-715-331
|
DuPont™
SL122BWH2X0012BN |
2X-Large |
Case of 12 for $728.00
|
|
|||||
19-715-329
|
DuPont™
SL122BWHMD0012BN |
Medium |
Case of 12 for $666.00
|
|
|||||
19-715-330
|
DuPont™
SL122BWHXL0012BN |
X-Large |
Case of 12 for $671.00
|
|
|||||
Description
Tychem™ 4000 delivers effective protection against a range of chemical environments. Tychem™ 4000, utilizes Saranex™ 23-P film laminated to Tyvek™ fabric to provide a rugged and durable fabric that resists abrasion. Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Attached hood with elastic around face opening
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
Compliance
BN=BERRY COMPLIANT
Specifications
White | |
Attached Socks | |
Hooded | |
Non-sterile |
Yes | |
Elastic | |
Bound |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.