Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DuPont™ Tychem™ 4000 Coverall with Standard Fit Hood, Attached Socks, Berry Compliant
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
Tychem™ 4000 delivers effective protection against a range of chemical environments. Tychem™ 4000, utilizes Saranex™ 23-P film laminated to Tyvek™ fabric to provide a rugged and durable fabric that resists abrasion. Tychem™ 4000 fabric provides at least 30 minutes of protection against >124 chemical challenges. Typical applications: chemical mixing, hazardous remediation, emergency medical response, paint spraying, and nuclear environments.
- Tightly sewn seam is covered with garment fabric to reinforce seam and reduce potential for particle penetration
- Attached hood with elastic around face opening
- Storm flap covers zipper which can be sealed by the wearer with adhesive strip to prevent intrusion at zipper
- Elastic opening for tighter fit at wrist
- Integrated socks composed of garment material
Compliance
BN=BERRY COMPLIANT
Specifications
Specifications
Size | 4X-Large |
Gender | Unisex |
Color | White |
Disposable | Yes |
Ankle Style | Attached Socks |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Storm flap with adhesive closure |
Seam Type | Bound |
Sterility | Non-sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Share a Content Correction