Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-1 Packed Column Ferrule, 1/8 in.
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only. Material Composition: Ceramic-filled PTFE. Maximum Temperature: 250°C
$402.00
Specifications
Description | Ferrule Type: Packed Column |
---|---|
Diameter (English) Outer | 1/8 in. |
Diameter (English) Inner | 1/8 in. |
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Packaging Quantity | Price | Quantity & Availability | |||||
11-100-9471
|
MilliporeSigma™ Supelco™
22309 |
100/Pk. |
Each for $402.00
|
|
|||||
11-101-1805
|
MilliporeSigma™ Supelco™
22496 |
10/Pk. | N/A | N/A | N/A | ||||
Specifications
Ferrule Type: Packed Column | |
1/8 in. | |
1/8 in. |
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC