Learn More
MilliporeSigma™ Supelco™ Supeltex™ M-1 Packed Column Ferrule, 1/8 in.
Characteristics: Ideal for connection to mass spectrometers. High reusability. Isothermal use only. Material Composition: Ceramic-filled PTFE. Maximum Temperature: 250°C
Supplier:  MilliporeSigma™ Supelco™ 22496
 View more versions of this product
Catalog No. 11-101-1805
Description
Description
Specifications
Specifications
1/8 in.
1/8 in.
10/Pk.
Ferrule Type: Packed Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
Supeltex is a registered trademark of Sigma-Aldrich Co. LLC