Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ SUPELCOSIL™ LC-ABZ Supelguard Cartridge, 5 μm Particle Size
SUPELCOSIL™ LC-ABZ phase has a unique deactivation technology which provides excellent reversed-phase performance for basic compounds, as well as those that are acidic, polar neutral, and non-polar.
Supplier: MilliporeSigma™ Supelco™ 59545U
Description
Silanol suppressing practices such as using competing amines, operating at high ionic strength, and adjusting mobile phases to pH extremes are typically not required to obtain good peak shape on the LC-ABZ phase.Specifications
2/Pk. | |
2 cm | |
Guard Cartridge | |
Reversed Phase | |
SUPELCOSIL Supelguard |
5 μm | |
120 Å | |
4 mm | |
HPLC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC
Spot an opportunity for improvement?Share a Content Correction