Learn More
Restek™ Rxi™-1301Sil MS Capillary Columns
SDP
Columns with the highest thermal stability in the industry ensure dependable, accurate MS results and increased uptime
Supplier:  Restek™ 16107
 View more versions of this product
Catalog No. 06-611-075
Explore all special offers
Add to Cart
Description
Description
  • Stabilized cyano phase selectivity improves the performance of existing methods
  • Rigorous QC testing ensures inertness and accurate, reliable data for multiple compound classes
  • Ideal for solvents, glycols, and other polar compounds
  • Maximum temperature: 320°C
Specifications
Specifications
Rxi™-1301Sil MS
0.53 mm
3 μm
Midpolar
1/Pk.
320°C
Solvents, Glycols, and Other Polar Compounds Analysis
60 m
Fused Silica
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.