Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Rxi™-1301Sil MS Capillary Columns
SDP

Columns with the highest thermal stability in the industry ensure dependable, accurate MS results and increased uptime

Supplier:  Restek™ 16104

 View more versions of this product

Catalog No. 06-611-072


Explore all special offers
Add to Cart

Description

Description

  • Stabilized cyano phase selectivity improves the performance of existing methods
  • Rigorous QC testing ensures inertness and accurate, reliable data for multiple compound classes
  • Ideal for solvents, glycols, and other polar compounds
  • Maximum temperature: 320°C
Specifications

Specifications

320°C
0.32 mm
1.5 μm
Midpolar
1/Pk.
Rxi™-1301Sil MS
Solvents, Glycols, and Other Polar Compounds Analysis
30 m
Fused Silica
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.