Learn More
Tocris Bioscience™ Rhosin hydrochloride
Rho GTPase inhibitor
$651.92
Specifications
Purity >98%
Inhibitors Rho GTPase Inhibitors
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-035-0R
View Documents
Tocris Bioscience™
5003/50
50 mg
Each for $651.92
Add to Cart
50-031-0R
View Documents
Tocris Bioscience™
5003/10
10 mg N/A N/A N/A
Description
Description
Rho GTPase inhibitor (Kd = 354 nM). Inhibits binding of RhoA to guanine nucleotide exchange factors (GEFs) including LARG, DBL, LBC, p115 RhoGEF and PDZ RhoGEF. Has no effect on the binding of Rac1 or Cdc42 to their respective GEFs. Suppresses RhoA/C-mediated breast cancer cell migration and invasion in vitro. Also induces neurite outgrowth in synergy with NGF.
Specifications
Specifications
>98%
Rho GTPase Inhibitors
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.