Learn More
Thermo Scientific™ Rapid DNA Ligation Kit PROMO
Rapid DNA Ligation Kit enables fast sticky-end or blunt-end DNA ligation in only 5 minuntes at room temperature.
Supplier: Thermo Scientific™ K1423
Description
The kit contains T4 DNA ligase and a specially formulated 5X rapid ligation buffer optimized for fast and efficient DNA ligation. Fast ligation efficiency is equal to that obtained with T4 DNA ligase in a standard 1 hour ligation. The ligation reaction mixture can be used directly for bacterial transformation with the TransformAid Bacterial Transformation Kit or with other conventional transformation procedures.
- Fast–ligation of either sticky- or blunt-end DNA in only 5 minutes
- Convenient–reaction mixture can be used directly for bacterial transformation
Recommended for:
Routine cloning experiments; Blunt-end cloning; Library construction; TA cloning.
Note:
Lyophilized bacterial strains GM2163 (†M0099) and JM107 (†M0109) can be provided free of charge upon request, with any purchase. Prior to electroporation it is necessary to inactivate T4 DNA ligase (1) by chloroform extraction.
Specifications
Kit | |
DNA Ligase | |
Rapid DNA Ligation Kit |
DNA | |
Nucleic Acid Gel Electrophoresis | |
150 Reactions |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.