Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ TACS-XL In Situ Apoptosis Detection Kits
A TUNEL assay designed for fixed cells, embedded tissue, and frozen tissue using light and electron microscopy
$465.16 - $568.50
Specifications
Quantity | 30 Tests |
---|---|
Product Type | TACS-XL In Situ Apoptosis Detection Kit |
Includes: Proteinase K Solution; Cytonin; TdT enzyme, B-dNTP mix; Strep-HRP; anti-BrdU; Strep-Diluent
Catalog Number | Mfr. No. | Detection Method | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Detection Method | Price | Quantity & Availability | |||||
482830BK
|
R&D Systems™
482830BK |
Blue Label |
Each for $562.60
|
|
|||||
482830DK
|
R&D Systems™
482830DK |
DAB |
Each for $568.50
|
|
|||||
482830K
|
R&D Systems™
482830K |
Basic |
Each for $465.16
|
|
|||||
482830R
|
R&D Systems™
482830R |
Replenisher | N/A | N/A | N/A | ||||
Description
Based on incorporation of bromodeoxyuridine (BrdU) at the 3 ft. OH ends of the DNA fragments that form during apoptosis
This detection system uses a biotin conjugated anti-BrdU antibody and streptavidin-horseradish peroxidase for visualiza
Specifications
30 Tests | |
TACS-XL In Situ Apoptosis Detection Kit |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title