Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Proteome Purify 12 Human Serum Protein Immunodepletion Resin
A single-use alternative to existing column or spin cartridge systems to deplete serum or plasma of high abundance proteins
$554.00 - $1050.00
Specifications
Product Type | Immunodepletion Resin |
---|---|
Content And Storage | Store the unopened product at 2 - 8° C. Do not use past expiration date. |
Includes: Immunodepletion Resin, Spin-X Filter Units
Description
Depletes samples of the 12 high-abundance proteins comprising approximately 95% of proteins in human serum or plasma
Depletes samples of albumin, IgA, IgG and IgM, Fibrinogen, Transferrin, Haptoglobin and others
Removal allows visualization of proteins
Specifications
Immunodepletion Resin | |
Store the unopened product at 2 - 8° C. Do not use past expiration date. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title