Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Proteome Purify 12 Human Serum Protein Immunodepletion Resin

A single-use alternative to existing column or spin cartridge systems to deplete serum or plasma of high abundance proteins

Supplier:  R&D Systems™ IDR012020

 View more versions of this product

Catalog No. IDR012020


Explore all special offers
Add to Cart

Includes: Immunodepletion Resin, Spin-X Filter Units
This item is not returnable. View return policy

Description

Description

Depletes samples of the 12 high-abundance proteins comprising approximately 95% of proteins in human serum or plasma
Depletes samples of albumin, IgA, IgG and IgM, Fibrinogen, Transferrin, Haptoglobin and others
Removal allows visualization of proteins

Specifications

Specifications

20 Tests
Immunodepletion Resin
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.