Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ DOT Shipping Labels

For marking packages with hazardous materials

Specifications

Format Label
Product Type DOT Shipping Label
Width (English) 4 in.
Height (English) 4 in.
Height (Metric) 10.16 cm
View More Specs

Products 14
Catalog Number Mfr. No. Quantity Display Text Material Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Display Text Material Price Quantity & Availability  
17-981-56H
View Documents
Fisherbrand™
DL 12A P
25/Pk. Corrosives 8 Vinyl N/A N/A N/A
17-985-411F
View Documents
Fisherbrand™
DL15AP
25/Pk. Organic Peroxide Vinyl N/A N/A N/A
17-987-170B
View Documents
Fisherbrand™
DL 4AL
25/Pk. Flammable Liquid Vinyl N/A N/A N/A
17-981-56D
View Documents
Fisherbrand™
DL161AP
25/Pk. Flammable Liquid 3 Paper N/A N/A N/A
17-987-170C
View Documents
Fisherbrand™
DL 6AL
1 Ea. Non Flammable Gas Paper N/A N/A N/A
17-987-170F
View Documents
N/A 500/Pk. Flammable Solid Paper N/A N/A N/A
17-987-170G
View Documents
N/A 500/Pk. Corrosive Paper N/A N/A N/A
17-981-56G
View Documents
N/A 25/Pk. Flammable Solids Vinyl N/A N/A N/A
17-981-56J
View Documents
N/A 25/Pk. Oxidizers And organic peroxides Vinyl N/A N/A N/A
17-987-170L
View Documents
N/A 500/Pk. Radioactive I Paper N/A N/A N/A
17-987-170A
View Documents
N/A 500/Pk. Flammable Gas Paper N/A N/A N/A
17-987-170M
View Documents
N/A 500/Pk. Radioactive II Paper N/A N/A N/A
17-981-56C
View Documents
N/A 25/Pk. Corrosive Vinyl N/A N/A N/A
17-981-56F
View Documents
N/A 25/Pk. Non-flammable Gas 2 Vinyl N/A N/A N/A
Description

Description

  • Help to meet the requirements in 49CFR for P.O.P. (“Performance Oriented Packaging”) labeling of hazardous material shipments
  • Must be retained on packaging until it is sufficiently cleaned of residue and purged of vapors

Order Info

Measure 4 x 4 in. . Availalble in pressure-sensitive paper or vinyl.

Specifications

Specifications

Label
4 in.
10.16 cm
DOT Shipping Label
4 in.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.