Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ DOT Shipping Labels
For marking packages with hazardous materials
Supplier: Fisherbrand™ DL15AP
Description
- Help to meet the requirements in 49CFR for P.O.P. (“Performance Oriented Packaging”) labeling of hazardous material shipments
- Must be retained on packaging until it is sufficiently cleaned of residue and purged of vapors
Order Info
Measure 4 x 4 in. . Availalble in pressure-sensitive paper or vinyl.
Specifications
DOT 49 CFR 172.519 | |
Vinyl | |
Organic Peroxide | |
25/Pk. | |
4 in. |
Label | |
DOT Shipping Label | |
Apply proper labels onto the outside of packing containers indentifying the appropriate hazard | |
4 in. | |
10.16 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction