Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthol AS-TR Phosphate 95.0+%, TCI America™
SDP

Catalog No. C2250200MG
Click to view available options
Quantity:
200 mg

Chemical Identifiers

CAS 2616-72-0
Molecular Formula C18H15ClNO5P
Molecular Weight (g/mol) 391.74
MDL Number MFCD00042716
InChI Key SHTFMUPUBYXPCD-UHFFFAOYSA-N
Synonym 3-[(4-Chloro-o-tolyl)carbamoyl]-2-naphthylphosphoric Acid
PubChem CID 75794
IUPAC Name ({3-[(4-chloro-2-methylphenyl)carbamoyl]naphthalen-2-yl}oxy)phosphonic acid
SMILES CC1=CC(Cl)=CC=C1NC(=O)C1=C(OP(O)(O)=O)C=C2C=CC=CC2=C1

Specifications

Quantity 200 mg
Formula Weight 391.74
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Naphthol AS-TR Phosphate [for Biochemical Research]
TSCA No
Recommended Storage Freezer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.