Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Pokeweed Mitogen, Lyoph.

Phytolacca Americana

$99.00 - $302.00

Specifications

For Use With (Application) Immunoassay
pH Range 7.2 to 7.4
Storage Requirements 2° to 8°C
Recombinant Recombinant
Form Lyophilized
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP08780332
View Documents
MP Biomedicals™
08780332
1mg
Each for $99.00
Add to Cart
 
ICN780331
View Documents
MP Biomedicals™
08780331
5mg
Each for $302.00
Add to Cart
 
Description

Description

Specifications

Specifications

Immunoassay
2° to 8°C
Lyophilized
7.2 to 7.4
Recombinant
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.