Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Pokeweed Mitogen, Lyoph.
Phytolacca Americana
Supplier: MP Biomedicals™ 08780331
Specifications
Immunoassay | |
7.2 to 7.4 | |
2° to 8°C | |
Lyophilized |
Pokeweed Mitogen, (Phytolacca Americana) from Phytolacca Americana | |
5mg | |
Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction