Learn More
Invitrogen™ IFN gamma Mouse ELISA Kit, Extra Sensitive
For the quantitative detection of mouse IFN-gamma.
$558.00 - $3910.00
Specifications
Accession Number | P01580 |
---|---|
Assay Range | 0.63-40 pg/mL |
Assay Sensitivity | 0.09 pg/mL |
Sample Type | Plasma, Serum, Supernatant |
Target Species | Mouse |
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to mouse IFN-gamma
Biotin-Conjugate anti-mouse IFN-gamma monoclonal antibody
Streptavidin-HRP
Mouse IFN-gamma Standard lyophilized, 80pg/mL upon reconstitution
Sample Diluent
Calibrator Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-246-677
|
Invitrogen™
BMS609 |
96 Tests |
Each for $558.00
|
|
|||||
50-246-678
|
Invitrogen™
BMS609TEN |
10 x 96 Tests |
Each for $3,910.00
|
|
|||||
Description
The Mouse Interferon gamma, extra sensitive (Ms IFNγ) ELISA quantitates Ms IFNγ in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IFNγ. Principle of the method The Mouse IFNγ solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistencySpecifications
P01580 | |
0.09 pg/mL | |
Mouse | |
Colorimetric Microplate Reader | |
IFN-g,Ifg | |
3.5% | |
HRP | |
Plasma, 20 μL; Serum, 20 μL; Supernatant, 20 μL | |
Immune interferon, IFG | |
1 hr. 20 min. |
0.63-40 pg/mL | |
Plasma, Serum, Supernatant | |
ELISA | |
15978 | |
2.7% | |
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Controls, Adhesive Plate Covers | |
RUO | |
2°C to 8°C | |
Mouse | |
3 hr. 30 min. |
For Research Use Only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.