Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MIP-2 (CXCL2) Mouse ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Supplier: Invitrogen™ 900K152
Description
Produced by activated monocytes and neutrophils and expressed at sites of inflammation. Hematoregulatory chemokine, which, in vitro, suppresses hematopoietic progenitor cell proliferation. GRO-beta(5-73) shows a highly enhanced hematopoietic activity.Specifications
P10889 | |
Lot-specific | |
ELISA | |
20310 | |
This Murine MIP-2 (CXCL2) ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate. | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hr 45 min |
Lot-specific | |
Biotin | |
Colorimetric Microplate Reader | |
Cxcl2 | |
HRP | |
RUO | |
Mouse | |
24 hr 30 min |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction