Learn More
Restek™ Micropacked Inlet Conversion Kits
SDP
Convert a Capillary GC Split/Splitless Inlet for use with 1/16 in. O.D. Micropacked Columns
$107.00 - $214.00
Specifications
Functionality/Interaction Type Sample pathways deactivated for ultimate inertness
View More Specs
Products 5
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
06-611-213
SDP
View Documents
Restek™
22428
Each for $107.00
Add to Cart
06-611-212
SDP
View Documents
Restek™
22427
Each for $164.00
Add to Cart
06-611-210
SDP
View Documents
Restek™
22425
Micropacked Column FID and Injection Port Adaptor Kit for On-Column Injection
Each for $214.00
Add to Cart
06-611-211
SDP
View Documents
Restek™
22426
Each for $122.00
Add to Cart
06-611-209
SDP
View Documents
Restek™
22424
Each for $212.00
Add to Cart
Description
Description
  • For use with Agilent 5890, 6890, and 7890 GCs
  • Sample pathways deactivated for ultimate inertness
Specifications
Specifications
Sample pathways deactivated for ultimate inertness
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.