Learn More
Restek™ Micropacked Inlet Conversion Kits
Convert a Capillary GC Split/Splitless Inlet for use with 1/16 in. O.D. Micropacked Columns
Supplier: Restek™ 22427
Description
- For use with Agilent 5890, 6890, and 7890 GCs
- Sample pathways deactivated for ultimate inertness
Specifications
Micropacked Column Injection Port Adaptor Kit | |
Sample pathways deactivated for ultimate inertness | |
On-Column Injection | |
Dual Vespel Ring Inlet Seal (Large Bore), Reducing Nut (Large Bore), 1/16 in. Ferrule (Vespel/Graphite), Siltek™ Treated Metal Liner Installation Guide, 1/16 in. Nut (Stainless Steel) |