Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ LumiBlue (TM) ECL Extended
$687.00 - $1086.00
Specifications
For Use With (Application) | Western Blot |
---|---|
Content And Storage | Store at 4°C |
Spot an opportunity for improvement?
Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title