Learn More
Anton Paar Set of 2 stainless steel jars 25 ml
Supplier:  Anton Paar 177680
Encompass_Preferred
Grinding jar with 25 mL volume made of INOX. The perfect solution for materials like minerals, cement, polymers, ores, etc.Suitable for use with Anton Paar BM500 Ball Mill NC1818284
Catalog No. NC1818288
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.