Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-4 Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Supplier: Invitrogen™ 900K14
This item is not returnable.
View return policy
Description
Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.Specifications
P05112 | |
Lot-specific | |
ELISA | |
3565 | |
This Human IL-4 ELISA Kit includes: Capture Antibody, Detection Antibody, Protein Standard, and HRP-Conjugate. | |
10 x 96 Tests | |
Refer to product documentation for component specific storage temperature. | |
1 hr 45 min |
Lot-specific | |
Biotin | |
Colorimetric Microplate Reader | |
Il4 | |
HRP | |
RUO | |
Human | |
24 hr 30 min |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction