Specifications
Empty Storm Case | |
HPX High Performance Resin | |
16.2 x 12.7 x 6.6 in. (41.1 x 32.26 x 16.76 cm) | |
6 in. | |
15.24 cm | |
16.2 in. | |
41.1 cm | |
6 lb. | |
12.7 in. | |
32.26 cm |
Orange | |
2 press and pull latches, 2 padlockable hasps, flush hinges | |
6.6 in. | |
16.76 cm | |
Double-layered, soft-grip handle, vortex valve | |
15 in. | |
38.1 cm | |
2.72 kg | |
10.5 in. | |
26.6 cm |