Learn More
Hardigg™ iM2200 Storm Cases
Supplier:  Hardigg™ IM220030001
 View more versions of this product
Catalog No. 19-130-5120
Description
Description
Specifications
Specifications
Divider Storm Case
HPX High Performance Resin
16.2 x 12.7 x 6.6 in. (41.1 x 32.26 x 16.76 cm)
6 in.
15.24 cm
16.2 in.
41.1 cm
6 lb.
12.7 in.
32.26 cm
Olive Drab
2 press and pull latches, 2 padlockable hasps, flush hinges
6.6 in.
16.76 cm
Double-layered, soft-grip handle, vortex valve
15 in.
38.1 cm
2.72 kg
10.5 in.
26.6 cm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.