Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hanks' Balanced Salts, Without Sodium Bicarbonate, MP Biomedicals™

Hanks' Balanced Salts, Without Sodium Bicarbonate, Orange Powder

$73.25

Specifications

Color Orange
pH 6.0 to 7.0 (before adjustment with sodium bicarbonate)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP091710122
View Documents
MP Biomedicals, Inc
091710122
1 x 10L
Each of 1 for $73.25
Add to Cart
 
Description

Description

  • Hanks' Balanced Salts solution is used to maintain the pH and osmotic balance in the medium and to provide the cells with water and essential inorganic ions.
Specifications

Specifications

Orange
6.0 to 7.0 (before adjustment with sodium bicarbonate)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.