Learn More
Galectin 1 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
$270.00 - $1836.00
Specifications
Antigen | Galectin 1 |
---|---|
Concentration | 0.1-1.0 mg/mL |
Applications | ELISA, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
500P210100
|
Pepro Tech Inc
500P210100UG |
100 μg |
Each for $270.00
|
|
|||||
500P2101MG
|
Pepro Tech Inc
500P2101MG |
1 mg |
Each for $1,836.00
|
|
|||||
Description
AA Sequence of recombinant protein: ACGLVASNLN LKPGECLRVR GEVAPDAKSF VLNLGKDSNN LCLHFNPRFN AHGDANTIVC NSKDGGAWGT EQREAVFPFQ PGSVAEVCIT FDQANLTVKL PDGYEFKFPN RLNLEAINYM AADGDFKIKC VAFD. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Galectin-1. Anti-Human Galectin-1-specific antibody was purified by affinity chromatography employing an immobilized Human Galectin-1 matrix. Sandwich ELISA: To detect Human Galectin-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human Galectin-1 (500-P210Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human Galectin-1. Western Blot: To detect Human Galectin-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human Galectin-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.Specifications
Galectin 1 | |
ELISA, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P09382 | |
3956 | |
E.coli-derived Recombinant Human Galectin-1 | |
Antigen affinity chromatography | |
LGALS1 |
0.1-1.0 mg/mL | |
Polyclonal | |
Lyophilized | |
RUO | |
PBS with no preservative | |
14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; beta-galactoside-binding lectin; beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; DKFZp686E23103; GAL1; GAL-1; Galaptin; Galbp; galectin 1; galectin 1; LGALS1; Galectin1; galectin-1; galectin-1-like protein; GBP; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; lectin, galactoside-binding, soluble, 1 (galectin 1); Lgals1; LGAS1; OPG-1; pm12; Putative MAPK-activating protein PM12; RL 14.5; similar to galactose-binding lectin; S-Lac lectin 1; unnamed protein product | |
LGALS1 | |
Primary | |
-20°C |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.