Learn More
Fisherbrand™ Universal All Purpose Absorbent Pillows
Designed to absorb almost any liquid
Description
Designed to absorb almost any liquid
- Designed for use in tight spaces, under valves or under machinery lifted off the floor, pillows provide the greatest absorbency per square inch
- Highly versatile, grey absorbents pick up water-based, petroleum-based and non-aggressive, chemical fluids
- Gray color masks spills and stains, helping you resist the urge to pick up the absorbent before it is fully saturated
- Made of all-purpose polypropylene suitable for most industrial applications
Compliance
OSHA 29 CFR 1910.22(a)(2); EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910,120(j)(1)(vii).
Specifications
15.4 gal. | |
58.2 L | |
22.8 x 22.8 cm (9 x 9 in.) | |
Polypropylene | |
9 in. | |
22.8 cm |
Gray | |
9 in. | |
22.8 cm | |
Universal All Purpose Absorbent Pillow | |
32/Cs. |