Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ELiTech Molecular Diagnostics Real Time PCR Positive Control Sets
Verify proper performance of an assay and detect problems during the DNA or RNA extraction and amplification process associated with real-time PCR assay
Specifications
Content And Storage | One vial of plasmid containing target insert associated with relevant ASR primers and ASR probes |
---|---|
No. of Tests | 24 |
For Use With (Application) | Real Time PCR (qPCR) |
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Qty | Availability | ||||
22-800-232
|
Nanogen, Inc
M400393 |
EBV | EBV | N/A | N/A | N/A | |||
22-800-231
|
Nanogen, Inc
M400394 |
CMV 3.0 | CMV 3.0 | N/A | N/A | N/A | |||
22-800-230
|
Nanogen, Inc
M400395 |
BK Virus | BK Virus | N/A | N/A | N/A | |||
22-800-238
|
Nanogen, Inc
M400396 |
VZV | VZV | N/A | N/A | N/A | |||
22-800-239
|
Nanogen, Inc
M400399 |
HSV-1 | HSV-1 | N/A | N/A | N/A | |||
22-800-233
|
Nanogen, Inc
M400401 |
HHV-6 | HHV-6 | N/A | N/A | N/A | |||
22-800-237
|
Nanogen, Inc
M400402 |
M. pneumonia | M. pneumonia | N/A | N/A | N/A | |||
22-800-236
|
Nanogen, Inc
M400406 |
C. pneumonia | C. pneumonia | N/A | N/A | N/A | |||
22-800-234
|
Nanogen, Inc
M400411 |
Enterovirus | Enterovirus | N/A | N/A | N/A | |||
22-800-235
|
Nanogen, Inc
M400412 |
hMPV | hMPV | N/A | N/A | N/A | |||
22-800-240
|
Nanogen, Inc
M400413 |
HSV-2 | HSV-2 | N/A | N/A | N/A | |||
Description
Each Positive Control ASR consists of plasmid containing the target insert associated with the relevant ASR primers and ASR probes product.
- Monitor aspects of PCR testing, lysis, extraction and PCR amplification, and detection
- Estimates testing precision and detects errors in laboratory testing procedures
- Effectively measures assay and instrument integrity
Specifications
One vial of plasmid containing target insert associated with relevant ASR primers and ASR probes | |
24 | |
Real Time PCR (qPCR) |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title