Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DHX38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

$535.76

Specifications

Antigen DHX38
Concentration 0.22 mg/mL
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-172-6021
SDP
Encompass_Preferred
View Documents
Proteintech Group Inc
100982AP150UL
150 μL
Each of 1 for $535.76
Add to Cart
 
Description

Description

Probable ATP-binding RNA helicase involved in pre-mRNA splicing.
Specifications

Specifications

DHX38
Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q92620
292007, 64340, 9785
DHX38 Fusion Protein Ag0144
Primary
-20°C
DHX38
0.22 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
DDX38, DEAH box protein 38, DHX38, KIAA0224, PRP16, PRPF16
Dhx38
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.