Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DHX38 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 100982AP150UL
Description
Probable ATP-binding RNA helicase involved in pre-mRNA splicing.Specifications
DHX38 | |
Polyclonal | |
Unconjugated | |
DHX38 | |
DDX38, DEAH box protein 38, DHX38, KIAA0224, PRP16, PRPF16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
292007, 64340, 9785 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q92620 | |
Dhx38 | |
DHX38 Fusion Protein Ag0144 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction