Learn More
R&D Systems™ Cultrex Collagen IV Cell Invasion Assay
Asseses cell invasion through Collagen IV
Supplier:  R&D Systems™ 3458024K
Catalog No. 34-580-24K
This item is not returnable. View return policy
Description
Description
  • Offers flexible utility, and sufficient insert size for informative results
  • Originally created in an effort to accelerate the screening process for compounds that influence cellular invasion through extracellular matrices, which is fundamental to angiogenesis, embryonic development, immune responses, and tumor cell metastasis.
  • Provides a platform for the analysis of responses to chemokines, toxins, drugs and other analytes of interest, for larger numbers of cells per well
  • Utilizes a simplified Boyden chamber design with an 8μm polycarbonate (PC) membrane
Specifications
Specifications
Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
24 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.