Learn More
METTLER TOLEDO™ Adapter Vials
Adapter accessories are made with conductive materials to reduce electrostatic charge build up. Our adapters load capsules, vials, bottles, or tubes in precisely the right position and secure them tightly in place for highly accurate dosing.
$450.00 - $719.00
Specifications
Type Adapter Vial
View More Specs
Products 33
Catalog Number Mfr. No. Size (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Size (Metric) Price Quantity & Availability  
01-676-972
View Documents
METTLER TOLEDO™
30459992
16.4 x 60 mm
Pack of 5 for $488.00
Add to Cart
01-676-973
View Documents
METTLER TOLEDO™
30459991
29.3 x 60 mm
Pack of 5 for $488.00
Add to Cart
01-676-975
View Documents
METTLER TOLEDO™
30459926
48 x 15 mm
Pack of 5 for $492.00
Add to Cart
01-676-977
View Documents
METTLER TOLEDO™
30459924
40.5 x 15 mm
Pack of 5 for $719.00
Add to Cart
01-676-978
View Documents
METTLER TOLEDO™
30459923
37.5 x 15 mm
Pack of 5 for $492.00
Add to Cart
01-676-979
View Documents
METTLER TOLEDO™
30459922
35.5 x 15 mm
Pack of 5 for $492.00
Add to Cart
01-676-980
View Documents
METTLER TOLEDO™
30459921
33.5 x 15 mm
Pack of 5 for $492.00
Add to Cart
01-676-981
View Documents
METTLER TOLEDO™
30428926
26 x 25 mm
Pack of 5 for $699.00
Add to Cart
01-676-982
View Documents
METTLER TOLEDO™
30428924
31.5 x 30 mm
Pack of 5 for $719.00
Add to Cart
01-676-983
View Documents
METTLER TOLEDO™
30428923
29.5 x 35 mm
Pack of 5 for $719.00
Add to Cart
01-676-984
View Documents
METTLER TOLEDO™
30428922
28.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-985
View Documents
METTLER TOLEDO™
30428921
27.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-986
View Documents
METTLER TOLEDO™
30428920
23.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-987
View Documents
METTLER TOLEDO™
30428919
22.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-988
View Documents
METTLER TOLEDO™
30428918
21.5 x 25 mm
Pack of 5 for $548.00
Add to Cart
01-676-989
View Documents
METTLER TOLEDO™
30428917
20.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-990
View Documents
METTLER TOLEDO™
30428916
19.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-991
View Documents
METTLER TOLEDO™
30428915
18.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-992
View Documents
METTLER TOLEDO™
30428914
17.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-993
View Documents
METTLER TOLEDO™
30428913
16.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-994
View Documents
METTLER TOLEDO™
30428912
15.5 x 35 mm
Pack of 5 for $492.00
Add to Cart
01-676-995
View Documents
METTLER TOLEDO™
30428911
14.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-676-996
View Documents
METTLER TOLEDO™
30428910
13.5 x 20 mm
Pack of 5 for $492.00
Add to Cart
01-676-997
View Documents
METTLER TOLEDO™
30428909
12.5 x 30 mm
Pack of 5 for $492.00
Add to Cart
01-676-998
View Documents
METTLER TOLEDO™
30428908
12 x 20 mm
Pack of 5 for $492.00
Add to Cart
01-676-999
View Documents
METTLER TOLEDO™
30428907
11.5 x 30 mm
Pack of 5 for $719.00
Add to Cart
01-677-000
View Documents
METTLER TOLEDO™
30428906
11 x 30 mm
Pack of 5 for $719.00
Add to Cart
01-677-001
View Documents
METTLER TOLEDO™
30428905
10.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-677-002
View Documents
METTLER TOLEDO™
30428904
10 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-677-003
View Documents
METTLER TOLEDO™
30428903
9.5 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-677-004
View Documents
METTLER TOLEDO™
30428902
9 x 25 mm
Pack of 5 for $492.00
Add to Cart
01-677-005
View Documents
METTLER TOLEDO™
30428901
8.5 x 15 mm
Pack of 5 for $450.00
Add to Cart
01-677-012
View Documents
METTLER TOLEDO™
30306209
1.5 mL
Pack of 5 for $488.00
Add to Cart
Description
Description
  • Secures vessel in precise position
  • Compatible with Q3 Sample Changer
  • Conductive materials to reduce electrostatic charge build-up
  • Highly accurate and safe filling
Specifications
Specifications
Adapter Vial
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.