Learn More
METTLER TOLEDO™ Adapter Vials
Adapter accessories are made with conductive materials to reduce electrostatic charge build up. Our adapters load capsules, vials, bottles, or tubes in precisely the right position and secure them tightly in place for highly accurate dosing.
Supplier:  METTLER TOLEDO™ 30428915
 View more versions of this product
Catalog No. 01-676-991
Explore all special offers
Add to Cart
Description
Description
  • Secures vessel in precise position
  • Compatible with Q3 Sample Changer
  • Conductive materials to reduce electrostatic charge build-up
  • Highly accurate and safe filling
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q3 Sample Changer (30418662)
18.5 x 25 mm
Adapter Vial
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.