Learn More
Enzo Life Sciences Zoledronic acid (25mg). CAS: 165800-06-6
SDP
Supplier:  Enzo Life Sciences ALX430153M025
Encompass_Preferred
Inhibitor of osteoclastic bone resorption. Prevents osteoporosis and skeletal fractures, particularly in patients with multiple myeloma or prostate cancer. Also used to treat hypercalcemia. Inhibition of acid sphingomyelinase (aSMase) with an IC50 5µm. Alternative name: 1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid, CGP 42446. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in 0.1N NaOH. Sparingly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-9240
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.