Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zobell’s Solution, Reagents
Zobell’s Solution, Reagent, use for Redox. Manufactured in ISO 9001 facility.
$74.25 - $319.00
Specifications
Chemical Name or Material | Zobell’s Solution |
---|---|
CAS | 7732-18-5, 7447-40-7, 14459-95-1 |
Packaging | Opaque HDPE |
Color | Yellow/Green |
For Use With (Equipment) | Redox Meter |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-333-368
|
Reagents Holdings Llc
CZ0001001B |
1 L |
Each for $107.00
|
|
|||||
01-333-367
|
Reagents Holdings Llc
CZ0001004B |
4 L |
Each for $319.00
|
|
|||||
01-333-366
|
Reagents Holdings Llc
CZ000100500B |
500 mL |
Each for $74.25
|
|
|||||
Specifications
Zobell’s Solution | |
Opaque HDPE | |
Redox Meter | |
Reagent | |
1.0048 | |
Completely miscible |
7732-18-5, 7447-40-7, 14459-95-1 | |
Yellow/Green | |
Oxidation Reduction Potential Standard | |
100°C | |
Redox | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title