Learn More
Enzo Life Sciences ZM 336372 (25mg). CAS: 208260-29-1
SDP
Supplier:  Enzo Life Sciences BMLEI2980025
Encompass_Preferred
Potent and specific inhibitor of the protein kinase c-Raf (IC50=70nM). Has no significant effect on many other protein kinases tested (even at 50µM) with the exception of p38 (SAPK2a) (IC50=2µM) and SAPK2b (p38β) (IC50=2µM). Induces a paradoxical >100-fold activation of c-Raf in whole cells. Alternative name: N-[5-(3-Dimethylaminobenzamido)-2-methylphenyl]-4-hydroxybenzamide. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO (200mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0683
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.