Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Zinc-Cast Slotted Masses Set
SCI_ED

Alternate for expensive brass weights.

Supplier:  Eisco™ PHZNW200SET

 View more versions of this product

Catalog No. S24449


Explore all special offers
Add to Cart

Description

Description

  • Zinc die-casted with powder coat
  • Set of nine weights and one hanger
  • Accuracy ±2%
Specifications

Specifications

Zinc Die-cast
200 g
Set of 9 x 20g weights and 1 x 20g hanger
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.