Learn More
Enzo Life Sciences Z-YVAD-pNA (1mg)
SDP
Supplier:  Enzo Life Sciences ALX260049M001
Encompass_Preferred
Chromogenic substrate for caspase-1 with increased cell permeability. λmax of pNA is 400nm. Alternative name: Caspase 1 substrate (chromogenic). Formula: C35H40N6O11. MW: 720.7. Purity: ≥97% (HPLC). Appearance: White powder. Formulation: Lyophilized. TFA salt. Solubility: Soluble in DMSO. Can be diluted with basic buffers. Long Term Storage: -20°C. Handling: Keep cool and dry.
Catalog No. 50-200-8263
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.