Learn More
Enzo Life Sciences Z-DEVD-pNA (1mg)
Supplier: Enzo Life Sciences ALX260048M001
Chromogenic substrate for caspase-3 with increased cell permeability. Also substrate for caspase-1, -4 and -7. λmax of pNA is 400nm. Similar to Ac-DEVD-pNA except the N-terminus is blocked by a benzyloxycarbonyl (Z) moiety which may increase cell permeability. Alternative name: Caspase 3 substrate (chromogenic). Formula: C32H38N6O14. MW: 730.7. Purity: ≥99% (HPLC). Formulation: Lyophilized. Solubility: Soluble in DMSO or acetonitrile. Long Term Storage: -20°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.