Learn More
Hach Company Radiometer Analytical XC161-9 Combination pH Electrode for Microsamples (d=5 mm, glass body, screw cap)
Supplier: Hach Company B10C161
The Radiometer XC161 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl) and micro diameter (5 mm) for small volume sample measurements. The XC161 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibility with Hach Sension+ laboratory pH meters.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.